Få mere at vide
SHP Steriltechnik AG SegoSoft™ Software
Artikelnummer. 31908789
Skift visning
Til brug med (udstyr):
Autoclaves
Artikelnummer. Til brug med (udstyr)
31908789 Autoclaves
1 options
Artikelnummer. 31908789 Leverandør SHP Steriltechnik AG Leverandørnr. 30.0000.600.000
 Begrænset vare
Forventet forsendelse: 14-10-2026
Læg i indkøbskurv
Læg i indkøbskurv
SegoSoft™ is a Windows‐based software solution designed for fully automatic digital documentation and traceability of instrument reprocessing workflows in medical and laboratory environments.
  • SegoSoft automatically captures and documents processing cycles from connected devices, including start/end times and all relevant cycle data
  • It supports over 200 different devices for sterilisation, cleaning and disinfection, so it works with equipment from a wide range of manufacturers
  • Documentation is stored in a format protected against unauthorized changes and can be archived long‐term for quality management and audit purposes
  • The software includes a unique user administration system based on digital signatures to ensure authenticated approvals and tamper‐proof records
  • It offers barcode label printing for easy traceability of batches and assignment to patient records, often via add‐ons like SegoLabel and SegoAssign
  • SegoSoft integrates into existing practice software systems and workflows, helping reduce manual documentation work and minimise liability risk
Tekniske data
Til brug med (udstyr) Autoclaves
Produkttype Software