Learn More
VITLAB™ Sedimentation Cone
Product Code. 11774129
Change view
Click to view available options
Volume (Metric):
1000 mL
Unit Size:
Pack of 3
Product Code. Volume (Metric) Unit Size
11774129 1000 mL Pack of 3
1 options
Product Code. 11774129 Supplier VITLAB™ Supplier No. 75991
  • ONLINE EXCLUSIVE PRICE
    850.00 DKK
    620.85 DKK / Pakke med 3
    Spare 229.15 DKK
    26% Slukket
Forventet forsendelse: 11-01-2027
Manufactured according to DIN 12 672. VITLAB™ Sedimentation Cone is used to determine suspended matter in liquids.
  • Clear
  • With raised scale 
  • Screw coupling can be removed
Order Info
CapacityMetric
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 1000 mL
Material SAN
No. per Pack 3
Type Sedimentation Cone