Få mere at vide
Securinine, TRC
Artikelnummer. 30134985
Skift visning
Mængde:
5 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134985 100 mg
30135470 50 mg
30010344 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134985 Leverandør Toronto Research Chemicals Leverandørnr. TRCS241000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5610-40-2
Molekylær formel C13 H15 N O2
Molekylvægt (g/mol) 217.26
SMIL O=C1O[C@@]23C[C@@H](C=CC2=C1)N4CCCC[C@H]34
Tekniske data
Kemisk navn eller materiale Securinine
Mængde 100 mg
InChI formel InChI=1S/C13H15NO2/c15-12-7-9-4-5-10-8-13(9,16-12)11-3-1-2-6-14(10)11/h4-5,7,10-11H,1-3,6,8H2/t10-,11-,13+/m1/s1
Formel vægt 217.1103