Learn More
3M™ Secure Click™ Particulate Filters
3M™ Secure Click™ Particulate Filter is our CE approved filter for environments containing certain particles.
Brand: 3M™ 7100197481
580.00 DKK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.21000kg
Description
- New dual flow cartridges and filters allow for easier breathing
- Protects against particulates with optional nuisance level organic vapour/acid gas relief
- Quick, simple connection. Simply align the connections and push until you hear a click
- Use with 3M™ Secure Click™ Reusable Respirators 800 Series
- Swept-back design allows an enhanced field of view and comfort
Specifications
CE Approved | |
P2R | |
D3125 |
Secure Click™ | |
Filter | |
80/Cs. |