Få mere at vide
Sartorius Secura™ Topload Weighing Balances: Promo
Shop alle Sartorius produkter
:
Weighing Range: 6100g; Readability; 10mg; Repeatability: 10mg; Linearity: 20mg
Beskrivelse
- Weighing Range: 6100g; Readability; 10mg; Repeatability: 10mg; Linearity: 20mg
- Promo offer expires 12-31-2016
Tekniske data
Tekniske data
| Volumen | 6.1 kg |
| Læsbarhed | 10 mg |
| Inkluderer | Universal power adapter, USB data cable, in-use co |
| Model- | 6102 |