Få mere at vide
Secorapamycin B, TRC
Artikelnummer. 30417415
Skift visning
Mængde:
1 mg
2.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30417415 1 mg
30414313 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30417415 Leverandør Toronto Research Chemicals Leverandørnr. TRCS2395551MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 185107-79-3
Molekylær formel C51H81NO14
Molekylvægt (g/mol) 932.19
Synonym [(2S,3E,5E,7E,9S,11R,13R,14R,15E,17R,20S,21R)-14,20-Dihydroxy-22-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-2,13-dimethoxy-3,9,11,15,17,21-hexamethyl-12,18-dioxo-3,5,7,15-docosatetraen-1-yl]tetrahydro-2-hydroxy-3-methyl-2H-pyran-2-yl]-2-oxoacetyl]-2-piperidinecarboxylic Acid, 34-Hydroxy Sirolimus, 34-Hydroxy Rapamycin
IUPAC navn (2R)-1-[2-[(2R,3R,6S)-6-[(2S,3E,5E,7E,13R,14R,15Z,21R)-14,20-dihydroxy-22-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-2,13-dimethoxy-3,9,11,15,17,21-hexamethyl-12,18-dioxodocosa-3,5,7,15-tetraenyl]-2-hydroxy-3-methyloxan-2-yl]-2-oxoacetyl]piperidine-2-carboxylic acid
SMIL CO[C@@H](C[C@@H]1CC[C@@H](C)[C@@](O)(O1)C(=O)C(=O)N2CCCC[C@@H]2C(=O)O)\C(=C\C=C\C=C\C(C)CC(C)C(=O)[C@H](OC)[C@H](O)\C(=C/C(C)C(=O)CC(O)[C@H](C)C[C@@H]3CC[C@@H](O)[C@@H](C3)OC)\C)\C
Tekniske data
Kemisk navn eller materiale Secorapamycin B
Mængde 1 mg
InChI formel InChI=1S/C51H81NO14/c1-30(24-34(5)45(56)47(65-10)46(57)35(6)25-32(3)41(54)29-42(55)33(4)26-37-20-22-40(53)44(27-37)64-9)16-12-11-13-17-31(2)43(63-8)28-38-21-19-36(7)51(62,66-38)48(58)49(59)52-23-15-14-18-39(52)50(60)61/h11-13,16-17,25,30,32-34,36-40,42-44,46-47,53,55,57,62H,14-15,18-24,26-29H2,1-10H3,(H,60,61)/b13-11+,16-12+,31-17+,35-25-/t30?,32?,33-,34?,36-,37+,38+,39-,40-,42?,43+,44-,46-,47+,51-/m1/s1
Formel vægt 931.5657