Få mere at vide
Secnidazole-d6, TRC
Artikelnummer. 30418327
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418327 50 mg
30418326 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418327 Leverandør Toronto Research Chemicals Leverandørnr. TRCS22500250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346603-27-7
Molekylær formel C7H5D6N3O3
Molekylvægt (g/mol) 191.22
Synonym α,2-Dimethyl-5-nitro-1H-imidazole-1-ethanol-d6, 1-(2-Methyl-5-nitroimidazol-1-yl) -2-propanol-d6, Flagentyl-d6, PM 185184-d6, RP 14539-d6
SMIL OC([2H])(C([2H])([2H])[2H])C([2H])([2H])N1C(C)=NC=C1[N+]([O-])=O
Tekniske data
Kemisk navn eller materiale Secnidazole-d6
Mængde 50 mg
InChI formel InChI=1S/C7H11N3O3/c1-5(11)4-9-6(2)8-3-7(9)10(12)13/h3,5,11H,4H2,1-2H3/i1D3,4D2,5D
Formel vægt 191.1177