missing translation for 'onlineSavingsMsg'
Få mere at vide

sec-Butyllithium, 1.3M in cyclohexane/hexane, packaged under Nitrogen in resealable AcroSeal™ bottles

Artikelnummer. 16675905
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16675905 100 mL
16665905 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Artikelnummer. 16675905 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H37922.AE
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 598-30-1 | C4H9Li | 64.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 598-30-1
Molekylær formel C4H9Li
Molekylvægt (g/mol) 64.06
MDL nummer MFCD00009323
InChI nøgle VATDYQWILMGLEW-UHFFFAOYNA-N
IUPAC navn (butan-2-yl)lithium
SMIL [Li]C(C)CC

Tekniske data

Mængde 100 mL
Fysisk form Liquid
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.