Få mere at vide
(±)-sec-Butyl-d9 Alcohol, CDN
Artikelnummer. 30321382
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321382 0.1 g 0.1g
30321370 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321382 Leverandør CDN Leverandørnr. CDND58410.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butan-2-ol-D9
CAS 1202864-22-9
Molekylær formel C4 D9 H O
Mængde 0.1 g
Synonym 2-Butan-1,1,1,2,3,3,4,4,4-d9-ol (ACI), sec-Butyl-d9 alcohol (6 CI), 1,1,1,2,3,3,4,4,4-Nonadeuteriobutan-2-ol, (±)-sec-Butyl-d9 alcohol, 2-Butanol-d9
InChI formel InChI=1 S/C4H10O/c1-3-4(2)5/h4-5 H,3H2,1-2H3/i1D3,2D3,3D2,4 D
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])(O)C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,2,3,3,4,4,4-nonadeuteriobutan-2-ol
Molekylvægt (g/mol) 83.1771
Formel vægt 83.1297 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity