Få mere at vide
(±)-sec-Butyl-3,3,4,4,4-d5 Alcohol, CDN
Artikelnummer. 30315629
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30315629 0.5 g 0.5g
30315321 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315629 Leverandør CDN Leverandørnr. CDND10630.5G
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Butan-3,3,4,4,4-d5-ol
CAS 75749-92-7
Molekylær formel C4 D5 H5 O
Mængde 0.5 g
Synonym 2-Butan-3,3,4,4,4-d5-ol (9 CI), sec-Butyl-d5 alcohol, 3,3,4,4,4-Pentadeuteriobutan-2-ol, (±)-sec-Butyl-3,3,4,4,4-d5 alcohol, 2-Butanol-d5, Butan-2-ol-3,3,4,4,4-D5
InChI formel InChI=1 S/C4H10O/c1-3-4(2)5/h4-5 H,3H2,1-2H3/i1D3,3D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C(C)O
IUPAC navn 3,3,4,4,4-pentadeuteriobutan-2-ol
Molekylvægt (g/mol) 79.1524
Formel vægt 79.1045 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity