Få mere at vide
(±)-sec-Butyl-1,1,1-d3 Alcohol, CDN
Artikelnummer. 30321476
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321476 0.25 g -
30310777 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321476 Leverandør CDN Leverandørnr. CDND10640.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Butanol-d3
CAS 53716-61-3
Molekylær formel C4 D3 H7 O
Mængde 0.25 g
Synonym sec-Butyl-2',2',2'-d3 Alcohol, 2-Butan-1,1,1-d3-ol, 2-Butan-1,1,1-d3-ol (9 CI), sec-Butyl-2',2',2'-d3 alcohol (6 CI), 1,1,1-Trideuteriobutan-2-ol, (±)-sec-Butyl-1,1,1-d3 alcohol, (±)-sec-Butyl-1,1,1-d3 alcohol, (±)-sec-Butyl-1,1,1-d3 alcohol
InChI formel InChI=1 S/C4H10O/c1-3-4(2)5/h4-5 H,3H2,1-2H3/i2D3
SMIL [2 H]C([2 H])([2 H])C(O)CC
IUPAC navn 1,1,1-trideuteriobutan-2-ol
Molekylvægt (g/mol) 77.14
Formel vægt 77.092 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity