Få mere at vide
Sebuthylazine-d5 (ethyl-d5), TRC
Artikelnummer. 30413341
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30413341 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30413341 Leverandør Toronto Research Chemicals Leverandørnr. TRCS2114215MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sebuthylazine D5
CAS 1219805-56-7
Molekylær formel C9 2H5 H11 Cl N5
Mængde 5 mg
Synonym Sebuthylazine D5 (N-ethyl D5), 6-Chloro-N4-(1,1,2,2,2-pentadeuterioethyl)-N2-sec-butyl-1,3,5-triazine-2,4-diamine
InChI formel InChI=1S/C9H16ClN5/c1-4-6(3)12-9-14-7(10)13-8(15-9)11-5-2/h6H,4-5H2,1-3H3,(H2,11,12,13,14,15)/i2D3,5D2
SMIL [2H]C([2H])([2H])C([2H])([2H])Nc1nc(Cl)nc(NC(C)CC)n1
IUPAC navn 2-N-butan-2-yl-6-chloro-4-N-(1,1,2,2,2-pentadeuterioethyl)-1,3,5-triazine-2,4-diamine
Molekylvægt (g/mol) 234.74
Formel vægt 234.1408