Learn More
Sampling Systems™ Sealing Sleeve Set
Sampling Systems™ Sealing Sleeve Set is designed to enable a pocket sampler to be emptied.
Brand: Sampling Systems™ 8005A-01
Additional Details : Weight : 0.00800kg
Description
- Enables one pocket to be emptied at a time
- Simply clip on to cover
- Easy to remove
Specifications
Polyethylene | |
Set of 3 sleeves for sampler | |
25 mm |