Learn More
Cytiva SE260/SE250 MiniVertical Electrophoresis System Replacement Parts: Spineless Combs
Brand: Cytiva 80614023
Additional Details : Weight : 0.00300kg
Specifications
SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit | |
PTFE | |
0.02 in. | |
9.5μL/ mm | |
0.03 in. |
Spineless Comb | |
5 | |
0.07 cm | |
1.3 cm | |
0.1 cm |