Learn More
Cytiva SE260/SE250 MiniVertical Electrophoresis System Replacement Parts: Spineless Combs
Brand:  Cytiva SE211A-10-1.0
689.87 DKK valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 0.45360kg
 View more versions of this product
Product Code. 10437694
  • 730.02 DKK / Each
Estimated Shipment: 21-05-2024
to see stock.
Specifications
SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit
PTFE
0.03 in.
4.8μL/ mm
0.03 in.
Spineless Comb
10
0.1 cm
0.48 cm
0.1 cm