Learn More
Cytiva SE260/SE250 MiniVertical Electrophoresis System Replacement Parts: Spineless Combs
Brand: Cytiva SE211A-15-1.5
Additional Details : Weight : 0.50000kg
Specifications
SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit | |
PTFE | |
0.05 in. | |
4.4μL/ mm | |
0.03 in. |
Spineless Comb | |
15 | |
0.15 cm | |
0.29 cm | |
0.1 cm |