Få mere at vide
SDZ 285-428, TRC
Artikelnummer. 30420570
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30420570 10 mg
30412218 5 mg
30418695 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30420570 Leverandør Toronto Research Chemicals Leverandørnr. TRCS21111310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 174262-13-6
Molekylær formel C24H20ClN3O
Molekylvægt (g/mol) 401.89
Synonym SDZ 285428, 4'-Chloro-N-[2-(1H-imidazol-1-yl)-2-phenylethyl][1,1'-biphenyl]-4-carboxamide
IUPAC navn N-(2-(1H-imidazol-1-yl)-2-phenylethyl)-4'-chloro-[1,1'-biphenyl]-4-carboxamide
SMIL O=C(C1=CC=C(C2=CC=C(Cl)C=C2)C=C1)NCC(N3C=NC=C3)C4=CC=CC=C4
Tekniske data
Kemisk navn eller materiale 4'-Chloro-N-[2-(1H-imidazol-1-yl)-2-phenylethyl][1,1'-biphenyl]-4-carboxamide
Mængde 10 mg
InChI formel InChI=1S/C24H20ClN3O/c25-22-12-10-19(11-13-22)18-6-8-21(9-7-18)24(29)27-16-23(28-15-14-26-17-28)20-4-2-1-3-5-20/h1-15,17,23H,16H2,(H,27,29)
Formel vægt 401.13