missing translation for 'onlineSavingsMsg'
Learn More
Scutellarin, MedChemExpress Product Code.: 16419088

Scutellarin, MedChemExpress

Product Code. p-200097864
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16419088

Brand: Medchem Express HYN0751

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Scutellarin, an active flavone isolated from Scutellaria baicalensis, can down-regulates the STAT3/Girdin/Akt signaling in HCC cells, and inhibits RANKL-mediated MAPK and NF-ÎşB signaling pathway in osteoclasts. Scutellarin is active against HIV-1IIIB, HIV-1(74V) and HIV-1KM018 with EC50s of 26 ÎĽM, 253 ÎĽM and 136 ÎĽM, respectively.

Scutellarin, an active flavone isolated from Scutellaria baicalensis, can down-regulates the STAT3/Girdin/Akt signaling in HCC cells, and inhibits RANKL-mediated MAPK and NF-ÎşB signaling pathway in osteoclasts. Scutellarin is active against HIV-1IIIB, HIV-1(74V) and HIV-1KM018 with EC50s of 26 ÎĽM, 253 ÎĽM and 136 ÎĽM, respectively.

Specifications

Chemical Name or Material Scutellarin
CAS 27740-01-8
Color Light Green
Purity Grade Notes Research
Molecular Formula C21H18O12
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (216.28 mM; Need ultrasonic)
SMILES O=C(C=C(C1=CC=C(O)C=C1)OC2=CC(O[C@@H]([C@@H]([C@@H](O)[C@@H]3O)O)O[C@@H]3C(O)=O)=C4O)C2=C4O
Molecular Weight (g/mol) 462.36
Formula Weight 462.36
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Scutellarin, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.