missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Scutellarin Product Code.: 16524474

Thermo Scientific Chemicals Scutellarin

Product Code. p-7214537 Shop All Thermo Scientific Chemicals Products
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 16524474

Brand: Thermo Scientific Chemicals 465132500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27740-01-8 | C21H18O12 | 462.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27740-01-8
Molecular Weight (g/mol) 462.36
InChI Key DJSISFGPUUYILV-ZFORQUDYSA-N
IUPAC Name (2S,3S,4S,5R,6S)-6-{[5,6-dihydroxy-2-(4-hydroxyphenyl)-4-oxo-4H-chromen-7-yl]oxy}-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES O[C@@H]1[C@@H](O)[C@H](OC2=C(O)C(O)=C3C(=O)C=C(OC3=C2)C2=CC=C(O)C=C2)O[C@@H]([C@H]1O)C(O)=O

Specifications

Molecular Formula C21H18O12
Quantity 250 mg
Formula Weight 462.36
Chemical Name or Material Scutellarin

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0463903 Certificate TypeCertificate of Analysis Date28/06/2024 Product Code465130010,465132500
Lot NumberA0413727 Certificate TypeCertificate of Analysis Date18/07/2022 Product Code465130010,465132500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Scutellarin >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.