missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ SCRIB Polyclonal Antibody

Description
Immunogen sequence: RLLEVNQQSLL GLTHGEAVQL LRSVGDTLTV LVCDGFEAST DAALEVSPGV IANPFAAGIG HRNSLESISS IDRE Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.
SCRIB is a cytoplasmic multimodular scaffold protein targeted to epithelial adherens junctions and neuronal presynaptic compartments. SCRIB and its orthologs in vertebrates and invertebrates participate in cell polarization.
Specifications
Specifications
| Antigen | SCRIB |
| Applikationer | Immunocytochemistry |
| Klassifikation | Polyclonal |
| Koncentration | 0.40 mg/mL |
| Konjugeret | Unconjugated |
| Formulering | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SCRIB |
| Genadgangsnr. | Q14160 |
| Gene Alias | AI118201; circletail; Crc; CRIB; CRIB1; hScrib; Kiaa0147; LAP4; mKIAA0147; PDZ-domain protein scribble; Protein LAP4; Protein scribble homolog; RGD1565055; S Vartul; SCRB1; Scrib; Scrib1; Scribble; scribble homolog 1; scribbled homolog; scribbled homolog (Drosophila); scribbled planar cell polarity protein; Vartul |
| Gensymboler | SCRIB |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?