Få mere at vide
Globe Scientific Skruelåg uden O-ring til prøverør
Artikelnummer. 31923809 Shop alle Globe Scientific produkter
Skift visning
Farve:
Blue
Gray
Green
Orange
Purple
Red
White
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Farve
31923809 Orange
31921971 Gray
31923729 Blue
31923054 Green
31923235 Purple
31922106 Red
31923193 White
31923158 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Artikelnummer. 31923809 Leverandør Globe Scientific Leverandørnr. 6136N
Forventet forsendelse: 11-11-2026
Læg i indkøbskurv
Læg i indkøbskurv
missing translation for 'validMainframeMsg'
Designet til opbevaring og transport af biologisk materiale, fremstillet af ikke-giftig polypropylen og tilbyder styrke og klarhed
  • The vial has external threads, which provide a smooth and uniform inner surface which reduces the risk of contamination
  • The screw caps are also made of polypropylene, providing the same coefficient of expansion as the tubes
  • This along with a polypropylene inner lip ensures a leakproof seal, even at changing temperatures
  • The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn
  • The caps are available with or with out a silicone o-ring and are available in a variety of colors. Screw caps sold separately
  • The screw caps are also made of polypropylene, providing the same coefficient of expansion as the tubes
  • This along with a polypropylene inner lip ensures a leakproof seal, even at changing temperatures
  • The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn
Tekniske data
Produkttype Screw Cap
Autoklaverbar Autoclavable
Materiale Polyethylene
Farve Orange
Sterilitet Non-sterile