Få mere at vide
Globe Scientific Skruelågsprøverør, ikke-graderede
Artikelnummer. 31923796 Shop alle Globe Scientific produkter
Skift visning
Bund form:
Conical (Self-standing)
Round
Round (Self-standing)
Kapacitet (metrisk):
1.2 mL
2 mL
3 mL
4 mL
5 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Bund form Kapacitet (metrisk)
31923796 Round 4 mL
31923641 Conical (Self-standing) 1.2 mL
31921818 Round 2 mL
31922475 Round (Self-standing) 2 mL
31922964 Round (Self-standing) 3 mL
31922344 Round (Self-standing) 4 mL
31922265 Round (Self-standing) 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Artikelnummer. 31923796 Leverandør Globe Scientific Leverandørnr. 6034
Forventet forsendelse: 11-11-2026
Disse rør og kapsler er designet til opbevaring og transport af biologisk materiale, med ikke-graderede materialer
  • Tubes are manufactured from non-toxic polypropylene and offer strength and clarity with non-graduated
  • The vial has external threads, which provide a smooth and uniform inner surface which reduces the risk of contamination
  • The screw caps are also made of polypropylene, providing the same coefficient of expansion as the tubes
  • This along with a polypropylene inner lip ensures a leakproof seal, even at changing temperatures
  • The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn
  • The caps are available with or with out a silicone o-ring and are available in a variety of colors
  • The screw caps are also made of polypropylene, providing the same coefficient of expansion as the tubes
  • This along with a polypropylene inner lip ensures a leakproof seal, even at changing temperatures
  • The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn
Tekniske data
Autoklaverbar Autoclavable
Kapacitet (metrisk) 4 mL
Bund form Round
Materiale Polypropylene
Trådstil External
Sterilitet Non-sterile
Uddannet No
Produkttype Sample Tube