Beskrivelse
- They are manufactured from non-toxic polypropylene and offer strength and clarity
- The vial has external threads, which provide a smooth and uniform inner surface which reduces the risk of contamination
- The screw caps are also made of polypropylene, providing the same coefficient of expansion as the tubes
- This along with a polypropylene inner lip ensures a leakproof seal, even at changing temperatures
- The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn
- The caps are available with or with out a silicone o-ring and are available in a variety of colors. Screw caps sold separately
- The screw caps are also made of polypropylene, providing the same coefficient of expansion as the tubes
- This along with a polypropylene inner lip ensures a leakproof seal, even at changing temperatures
- The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn
Tekniske data
Tekniske data
| Produkttype | Screw Cap with O-ring |
| Autoklaverbar | Autoclavable |
| Materiale | Polypropylene |
| Farve | Red |
| Sterilitet | Non-sterile |