Få mere at vide
Globe Scientific Skruedæksel med O-ring til prøverør
Artikelnummer. 31923913 Shop alle Globe Scientific produkter
Skift visning
Farve:
Blue
Clear
Gray
Green
Orange
Purple
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Farve
31923913 Red
31922169 Gray
31923813 Blue
31921919 Green
31923908 Purple
31922350 Orange
31923413 White
31922268 Yellow
31921542 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Artikelnummer. 31923913 Leverandør Globe Scientific Leverandørnr. 6137R
Forventet forsendelse: 11-11-2026
Disse rør og låg er designet til opbevaring og transport af biologisk materiale og giver styrke og klarhed
  • They are manufactured from non-toxic polypropylene and offer strength and clarity
  • The vial has external threads, which provide a smooth and uniform inner surface which reduces the risk of contamination
  • The screw caps are also made of polypropylene, providing the same coefficient of expansion as the tubes
  • This along with a polypropylene inner lip ensures a leakproof seal, even at changing temperatures
  • The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn
  • The caps are available with or with out a silicone o-ring and are available in a variety of colors. Screw caps sold separately
  • The screw caps are also made of polypropylene, providing the same coefficient of expansion as the tubes
  • This along with a polypropylene inner lip ensures a leakproof seal, even at changing temperatures
  • The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn
Tekniske data
Produkttype Screw Cap with O-ring
Autoklaverbar Autoclavable
Materiale Polypropylene
Farve Red
Sterilitet Non-sterile