Learn More
Lenz Laborglasintrumente Screw Caps
Click to view available options
Thread Size (Metric):
25 GL
32 GL
45 GL
Description
- Material: PBT (red)
- With PTFE-coated sealing
- Temperature resistance: up to 180 °C
Specifications
Color | Red |
Material | PBT |
Thread Size (Metric) | 32 GL |
Closure Material | PTFE |