missing translation for 'onlineSavingsMsg'
Få mere at vide

SCR7, MedChemExpress

Artikelnummer. 16488017
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16488017 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16488017 Leverandør Medchem Express Leverandørnr. HY12742

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress SCR7 is an unstable form that can be autocyclized into a stable form SCR7 pyrazine. SCR7 pyrazine is a DNA ligase IV inhibitor that blocks nonhomologous end-joining (NHEJ) in a ligase IV-dependent manner. SCR7 pyrazine is also a CRISPR/Cas9 enhancer which increases the efficiency of Cas9-mediated homology-directed repair (HDR). SCR7 pyrazine induces cell apoptosis and has anticancer activity.

SCR7 is an unstable form that can be autocyclized into a stable form SCR7 pyrazine. SCR7 pyrazine is a DNA ligase IV inhibitor that blocks nonhomologous end-joining (NHEJ) in a ligase IV-dependent manner. SCR7 pyrazine is also a CRISPR/Cas9 enhancer which increases the efficiency of Cas9-mediated homology-directed repair (HDR). SCR7 pyrazine induces cell apoptosis and has anticancer activity.

Tekniske data

Kemisk navn eller materiale SCR7
Fysisk form Powder
CAS 1533426-72-0
Farve Yellow
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C18H14N4OS
Til brug med (applikation) Cancer-Kinase/protease
Opløselighedsinformation DMSO : ≥ 45 mg/mL (134.57 mM)
SMIL O=C(C(/N=C/C1=CC=CC=C1)=C(/N=C/C2=CC=CC=C2)N3)NC3=S.[(E)].[(E)]
Molekylvægt (g/mol) 334.39
Formel vægt 334.39
Procent renhed 98.22%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.