missing translation for 'onlineSavingsMsg'
Få mere at vide

STEMCELL Technologies SCR7

Artikelnummer. 17100332
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17100332 10 mg
17190322 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17100332 Leverandør STEMCELL Technologies Leverandørnr. 74104

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

NHEJ pathway inhibitor; inhibits DNA ligase IV

  • SCR7 is an inhibitor of DNA ligase IV, which is responsible for the repair of DNA double-strandbreaks via the non-homologous end joining (NHEJ) repair pathway (Srivastava et al.) Due to its reported success in impeding cancer cell growth and potential impact on future cancer therapeutics, SCR7 has been closely studied in many recent publications (Hosoya & Miyagawa; John et al.)
  • Inhibits NHEJ-dependent DNA repair; this inhibition is reported to enhance precise homology-directed repair (HDR)-dependent CRISPR-Cas9 genome editing (Chu et al.; Maruyama et al.; Pinder et al.) However, these effects are cell type-specific and context-dependent (Song et al.; Xie et al.; Yang et al.; Zhang et al.)
  • Activates apoptosis of cancer cells by inhibiting DNA ligase IV to increase the efficacy of DNA double-strand break-inducing therapy (chemo- or radio-therapy) (Srivastava et al.)

Tekniske data

CAS 1533426-72-0
Mængde 10 mg
Molekylær formel C18H14N4OS
Renhed ≥95%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.