Beskrivelse
- Reseal and package heavy weights of valuable goods
- Reliable box sealing tape for medium- to heavy-weight packaging applications
- High initial tack and shear adhesion to fiberboard
- Use for single strip closure of fiberboard boxes
- Performs well at all temperatures typically encountered by packaged products in shipping and storage environments
- Quality tape for closing boxes
- Tape resists splitting when subjected to rough handling and absorbs impact shock
Tekniske data
Tekniske data
| Klæbende type | Hot-melt |
| Farve | Brun |
| Format | Tape |
| Produkttype | Tape |
| Dimensioner (L x B) | 66 m x 50 mm |
| Længde (engelsk) | 72.18 yd. |
| Bredde (engelsk) | 2 in. |