Få mere at vide
Scopolamine N-Oxide, TRC
Artikelnummer. 30005098
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005098 10 mg
30134289 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30005098 Leverandør Toronto Research Chemicals Leverandørnr. TRCS20002010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 97-75-6
Molekylær formel C17H21NO5
Molekylvægt (g/mol) 319.35
Synonym (αS)-α-(Hydroxymethyl)benzeneacetic Acid (1α,2β,4β,5α,7β)-9-Methyl-9-oxido-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl Ester, 6β,7β-Epoxy-1αH,5αH-tropan-3α-ol (-)-Tropate 8-Oxide, [7(S)-(1α,2β,4β,5α,7β)]-α-(Hydroxymethyl)benzeneacetic Acid 9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl Ester N-Oxide, Scopolamine, N-Oxide, 3-Oxa-9-azatricyclo[3.3.1.02,4]nonane Benzeneacetic Acid Deriv., Genoscopolamine, Hyoscinamine Oxide, Hyoscine N-Oxide, Hyoscineamine Oxide, Oxicopolamina, Scolamine, Scolen, Scopodex, Scopolamine aminoxide, Seamalin, Seronex
IUPAC navn [(1R,2R,4S,5S)-9-methyl-9-oxido-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonan-7-yl] (2S)-3-hydroxy-2-phenylpropanoate
SMIL O=C([C@H](CO)C1=CC=CC=C1)O[C@H]2C[C@H]3[C@H](O4)[C@H]4[C@@H](C2)N3(C)=O
Tekniske data
Kemisk navn eller materiale Scopolamine N-Oxide
Mængde 10 mg
InChI formel InChI=1S/C17H21NO5/c1-18(21)13-7-11(8-14(18)16-15(13)23-16)22-17(20)12(9-19)10-5-3-2-4-6-10/h2-6,11-16,19H,7-9H2,1H3/t11-,12-,13-,14+,15-,16+,18+/m1/s1
Formel vægt 319.142