Få mere at vide
Scopine, Mikromol™
Artikelnummer. 30050065
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050065 100 mg
30051761 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050065 Leverandør Mikromol Leverandørnr. MM0546.09
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Scopine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Scopine
CAS 498-45-3
Analyt- eller komponentnavne Scopine
Forsendelsestilstand Room Temperature
Molekylær formel C8 H13 N O2
Certificeringer/compliance ISO 9001
Synonym (1α,2β,4β,5α,7β)-9-methyl-3-Oxa-9-azatricyclo[3.3.1.02,4]nonan-7-ol, 1αH,5αH-Tropan-3α-ol, 6β,7β-epoxy- (8CI), Scopine (6CI,7CI), (1α,2β,4β,5α,7β)-9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-7-ol, 6,7-Epoxytropine, Scopanol, Scopin
InChI formel InChI=1S/C8H13NO2/c1-9-5-2-4(10)3-6(9)8-7(5)11-8/h4-8,10H,2-3H2,1H3/t4-,5-,6+,7-,8+
SMIL CN1[C@@H]2C[C@@H](O)C[C@H]1[C@@H]3O[C@H]23
IUPAC navn (1α,2β,4β,5α,7β)-9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-7-ol
Molekylvægt (g/mol) 155.19
Mængde 100 mg
Formel vægt 155.0946 g/mol