Få mere at vide
Scopine Di(2-thienylglycolate)-D3, TRC
Artikelnummer. 30409470
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30409470 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30409470 Leverandør Toronto Research Chemicals Leverandørnr. TRCS2001031MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Scopine Di(2-thienylglycolate)-D3
CAS 1126775-83-4
Molekylær formel C18H16D3NO4S2
Mængde 1 mg
Synonym N-Demethyl Tiotropium-D3, α-Hydroxy-α-2-thienyl-2-thiopheneacetic Acid (1α,2β,4β,5α,7β)-9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl Ester-D3, Tiotropium EP Impurity B-D3
InChI formel InChI=1S/C18H19NO4S2/c1-19-11-8-10(9-12(19)16-15(11)23-16)22-17(20)18(21,13-4-2-6-24-13)14-5-3-7-25-14/h2-7,10-12,15-16,21H,8-9H2,1H3/t10?,11-,12+,15-,16?/m1/s1/i1D3
SMIL OC(C1=CC=CS1)(C(O[C@@H]2C[C@H]3N(C([2H])([2H])[2H])[C@H](C4[C@@H]3O4)C2)=O)C5=CC=CS5
Molekylvægt (g/mol) 380.5
Formel vægt 380.0944