Få mere at vide
Scoparone, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Scoparone |
| CAS | 120-08-1 |
| Molekylær formel | C11 H10 O4 |
| Mængde | 500 mg |
| Synonym | 2H-1-Benzopyran-2-one','6,7-dimethoxy-, Coumarin','6,7-dimethoxy- (6CI,7CI,8CI), 6,7-Dimethoxy-2H-1-benzopyran-2-one, 6,7-Dimethoxy-2H-chromen-2-one, 6,7-Dimethoxycoumarin, 6,7-Dimethylesculetin, Aesculetin dimethyl ether, Escoparone, Esculetin 6,7-dimethyl ether, Esculetin dimethyl ether, NRB 03190, O,O-Dimethylesculetin, O-Methylisoscopoletin, O-Methylscopoletin, Scoparon, Scoparone, Scopoletin methyl ether, Scopoletin monomethyl ether |
| InChI formel | InChI=1S/C11H10O4/c1-13-9-5-7-3-4-11(12)15-8(7)6-10(9)14-2/h3-6H,1-2H3 |
| SMIL | COc1cc2OC(=O)C=Cc2cc1OC |
| IUPAC navn | 6,7-dimethoxychromen-2-one |
| Molekylvægt (g/mol) | 206.19 |
| Formel vægt | 206.0579 |