Få mere at vide
Scleroglucan (Technical Grade), TRC
Artikelnummer. 30415991
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30415991 500 mg
30412118 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30415991 Leverandør Toronto Research Chemicals Leverandørnr. TRCS200073500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Scleroglucan
CAS 39464-87-4
Molekylær formel C24H40O21
Mængde 500 mg
Synonym Actigum C 56DF, Actigum CS 11, Actigum CS 6, Actigum CS 6DF, Actigum CS II-L, Amigel, Amigum, Betasizofiran, Biovis, CS 11, Clearogel CS 11, Clearogel CS 11D, PM-II, Polytetran, Polytran FS, Scleroglucan gum, Sclerogum, Sclerosan, Sclerotan, Sclerotium gum, Sclerotium rolfsii gum
InChI formel InChI=1S/C24H44O20P2/c25-1-5-9(28)13(32)14(33)21(38-5)37-4-8-12(31)18(42-23-16(35)20(44-46)11(30)6(2-26)39-23)15(34)22(40-8)43-19-10(29)7(3-27)41-24(45)17(19)36/h5-36H,1-4,45-46H2
SMIL C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3C(C(OC(C3O)P)CO)O)O)OC4C(C(C(C(O4)CO)O)OP)O)O)O)O)O)O
IUPAC navn 2-[[6-[3,5-dihydroxy-2-(hydroxymethyl)-6-phosphanyloxan-4-yl]oxy-4-[3,5-dihydroxy-6-(hydroxymethyl)-4-phosphanyloxyoxan-2-yl]oxy-3,5-dihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Molekylvægt (g/mol) 664.56
Formel vægt 714.1901