Få mere at vide
SCH900776 (S-isomer), MedChemExpress
Artikelnummer. 16456796
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16456796 100 mg
16442697 50 mg
16426608 10 mg
16409177 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16456796 Leverandør Medchem Express Leverandørnr. HY15532B
MedChemExpress SCH900776 S-isomer is the S-isomer of SCH900776. SCH900776 is a potent, selective and orally bioavailable inhibitor of checkpoint kinase1 (Chk1) with IC50 of 3 nM.
SCH900776 S-isomer is the S-isomer of SCH900776. SCH900776 is a potent, selective and orally bioavailable inhibitor of checkpoint kinase1 (Chk1) with IC50 of 3 nM.
Tekniske data
Kemisk navn eller materiale SCH900776 (S-isomer)
Fysisk form Solid
CAS 891494-64-7
Farve Yellow
Mængde 100 mg
Renhedsgrad noter Research
Molekylær formel C15H18BrN7
Til brug med (applikation) Cancer-Kinase/protease
Synonym MK-8776 S-isomer
Opløselighedsinformation DMSO : ≥ 100 mg/mL (265.78 mM)
SMIL NC1=C(Br)C([C@@H]2CNCCC2)=NC3=C(C4=CN(C)N=C4)C=NN13
Molekylvægt (g/mol) 376.25
Formel vægt 376.25
Procent renhed 95.88%