missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SCH 79797 dihydrochloride

Description
Potent, selective non-peptide PAR1 receptor antagonist (IC50 = 70 nM). Inhibits haTRAP-induced- but not γ-thrombin-, ADP- or collagen-induced human platelet aggregation. Also selectively blocks PAR1 agonist- or thrombin-induced increases in cytosolic Ca2+ in vascular smooth muscle cells.
Specifications
Specifications
| Kemisk navn eller materiale | SCH 79797 dihydrochloride |
| Mængde | 50 mg |
| Formel vægt | 444.41 |
| Renhed | >99% |
| Molekylvægt (g/mol) | 444.404 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
¿Detecta una oportunidad de mejora?