Få mere at vide
SCH 336, Tocris Bioscience™
GREENER_CHOICE
Highly potent and selective CB2 inverse agonist
Brand:  Tocris Bioscience 5815/10
Encompass
Produktkode 15933231
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
SCH 336 is a highly potent and selective CB2 receptor inverse agonist (Ki = 1.8 nM, EC50 = 2 nM). Displays 100-fold selectivity for CB2 receptors over CB1. Increases forskolin stimulated cAMP accumulation in CHO cells expressing human CB2 receptors. Inhibits leukocyte migration in a murine model of delayed-type hypersensitivity and inhibits antigen-induced lung eosinophilia in a mouse allergy model.
Tekniske data
Tekniske data
N-[(1S)-1-[4-[[4-Methoxy-2-[(4-methoxyphenyl)sulfonyl]phenyl]sulfonyl]phenyl]ethyl]methanesulfonamide
447459-51-0
>98%
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner