Learn More
Buerkle™ Scrapers for Foodstuffs
Product Code. 10260512 Shop All Bürkle Products
Change view
Click to view available options
Length (Metric):
205 mm
250 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10260512 205 mm Each
10440401 250 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10260512 Supplier Buerkle™ Supplier No. 83004011
  • ONLINE EXCLUSIVE PRICE
    56.75 DKK
    54.11 DKK / Pakke med 1
    Spare 2.64 DKK
    4% Slukket
Forventet forsendelse: 05-01-2027
Scrapers with easy-to-clean, smooth and pore-free surface ideal for the food sector.
  • For foodstuffs (FDA and EU Nr. 10/2011)
  • Polypropylene has a high resistance to chemicals and temperatures
  • Can be also used for boiling products
TRUSTED_SUSTAINABILITY
Specifications
Product Type Scraper
Handle Type Fixed
Autoclavable Autoclavable
Certifications/Compliance FDA and (EU) Nr. 10/2011
Color White
For Use With (Application) Suitable for Use with Foodstuffs
Length (Metric) 205 mm
Material Polypropylene
Style One-piece
Width (Metric) 75 mm