missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAMP4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-81787
Additional Details : Weight : 0.00970kg
Description
SCAMP4 Polyclonal specifically detects SCAMP4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SCAMP4 | |
Polyclonal | |
Unconjugated | |
PBS | |
SCAMP4 | |
SCAMP4 antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human SCAMP4. The immunogen is located within the last 50 amino acids of SCAMP4. Amino Acid Squence: NSLPEYPTVPSYPGS | |
0.1 mg | |
Primary | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin | |
secretory carrier membrane protein 4 | |
Rabbit | |
Affinity Purified | |
RUO | |
113178 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |