missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ SBF1 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 16341895
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
Pakningsstørrelse:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16341895 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16341895 Leverandør Invitrogen™ Leverandørnr. PA584945

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Immunogen sequence: CYDSCPRAQP DAISRLLEEL QRLETELGQP AERWKDTWDR VKAAQRLEGR PDGRGTPSSL LVSTAPHHRR SLGVYL Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 89%.

Probable pseudophosphatase. Lacks several amino acids in the catalytic pocket which renders it catalytically inactive as a phosphatase. The pocket is however sufficiently preserved to bind phosphorylated substrates, and maybe protect them from phosphatases. Inhibits myoblast differentiation in vitro and induces oncogenic transformation in fibroblasts. According to PubMed:20937701, may function as a guanine nucleotide exchange factor (GEF) activating RAB28. Promotes the exchange of GDP to GTP, converting inactive GDP-bound Rab proteins into their active GTP-bound form.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen SBF1
Applikationer Immunocytochemistry
Klassifikation Polyclonal
Koncentration 0.5 mg/mL
Konjugeret Unconjugated
Formulering PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Sbf1
Genadgangsnr. O95248
Gene Alias CMT4B3; DENN/MADD domain containing 7A; DENND7A; Inactive phosphatidylinositol 3-phosphatase 5; MTMR5; Myotubularin-related protein 5; Sbf1; SET binding factor 1; SET-binding factor 1
Gensymboler Sbf1
Værtsarter Rabbit
Immunogen Recombinant protein corresponding to Human SBF1. Recombinant protein control fragment (Product #RP-108629).
Oprensningsmetode Antigen affinity chromatography
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 6305
Målarter Human
Indhold og opbevaring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.