missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
STEMCELL Technologies SB203580 (Hydrochloride)
Beskrivelse
- SB203580 (Hydrochloride) is a potent inhibitor of p38 MAPK activity (IC50 = 0.6 μM)
- It inhibits both the α and β isoforms of p38 MAPK, and does not inhibit ERK or JNK
- (Bain et al.), Cuenda et al.)
- Enhances the growth and self-renewal of mouse embryonic stem (ES) cells (Qi et al.)
- Promotes long-term maintenance of human naïve ground state pluripotent stem cells (Gafni et al.)
- Promotes proliferation of human endothelial progenitor cells (Seeger et al.)
- Promotes proliferation of neonatal and adult rat cardiomyocytes (Engel et al.)
- Enhances differentiation of cardiomyocytes from human ES cells (Gaur et al.), Graichen et al.)
- Inhibits differentiation of cardiomyocytes from mouse ES cells by inhibition of early mesoderm (Davidson and Morange)
Tekniske data
Tekniske data
| CAS | 869185-85-3 |
| Synonym | PB 203580; RWJ 64809 |
| Mængde | 5 mg |
| Molekylær formel | C21H16FN3OS · HCl |
| Renhed | ≥98% |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?