Få mere at vide
Tocris Bioscience™ SB 772077B dihydrochloride
GREENER_CHOICE
Artikelnummer. 18094142
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18094142 10 mg
18075222 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18094142 Leverandør Tocris Bioscience™ Leverandørnr. 4118/10
Potent Rho-kinase inhibitor; vasodilator
SB 772077B dihydrochloride is a potent Rho-kinase (ROCK) inhibitor (IC50 value of approximately 5.6 nM at recombinant human ROCK1 and 2). Decreases pulmonary and systemic arterial blood pressures and increases cardiac output. Exhibits vasodilator activity that is more potent than Y-27632 (Cat. No 1254) or Fasudil (Cat. No. 0541).
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 607373-46-6
Molekylær formel C15H20Cl2N8O2
Tekniske data
Kemisk navn eller materiale SB 772077B dihydrochloride
Mængde 10 mg
Formel vægt 415.28
Renhed >99%
Molekylvægt (g/mol) 415.279