Få mere at vide
Tocris Bioscience™ SB 706504
p38 MAPK inhibitor
Brand: Tocris Bioscience™ 5040/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SB 706504 is a p38 MAPK inhibitor. Inhibits LPS-induced transcription of a range of chemokines and cytokines in chronic obstructive pulmonary disease (COPD) monocyte derived macrophages (MDMs). Also inhibits LPS-induced protein expression of IL-6, IL-10, TNFα and γ-inducible protein 10 in COPD MDMs. Effects on suppression of LPS-induced gene and protein expression are enhanced in combination with dexamethasone (Cat. No. 1126).Tekniske data
| N-Cyano-N′-[2-[[8-(2,6-difluorophenyl)-4-(4-fluoro-2-methylphenyl)-7,8-dihydro-7-oxopyrido[2,3-d]pyrimidin-2-yl]amino]ethyl]guanidine | |
| 911110-38-8 | |
| 50 mg |