Få mere at vide
Tocris Bioscience™ SB 674042
GREENER_CHOICE
Artikelnummer. 30002039
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30002039 10 mg
30002047 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30002039 Leverandør Tocris Bioscience™ Leverandørnr. 4673/10
Potent and selective non-peptide OX1 antagonist
Potent and selective non-peptide orexin OX1 receptor antagonist (Kd = 3.76 nM). Exhibits 100-fold selectivity for OX1 over OX2 receptors. Has no effect at serotonergic, dopaminergic, adrenergic or purinergic receptors. Inhibits orexin 1-induced Ca2+ mobilization in CHO-DG44 cells stably transfected with the OX1 receptor.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 483313-22-0
Molekylær formel C24H21FN4O2S
Tekniske data
Kemisk navn eller materiale SB 674042
Mængde 10 mg
Formel vægt 448.51
Molekylvægt (g/mol) 448.516