missing translation for 'onlineSavingsMsg'
Få mere at vide

SB 525334, MedChemExpress

Artikelnummer. 16457687
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16457687 5 mg -
16454948 10 mg -
16421197 50 mg 50mg
16435726 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16457687 Leverandør Medchem Express Leverandørnr. HY12043

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress SB 525334 is a potent and selective transforming growth factor β1 receptor (ALK5) inhibitor with an IC50 of 14.3 nM.

SB 525334 is a potent and selective transforming growth factor β1 receptor (ALK5) inhibitor with an IC50 of 14.3 nM.

Tekniske data

Kemisk navn eller materiale SB 525334
CAS 356559-20-1
Farve Yellow
Renhedsgrad noter Research
Molekylær formel C21H21N5
Mængde 5 mg
Opløselighedsinformation DMSO : 100 mg/mL (291.19 mM; Need ultrasonic)
SMIL CC(C)(C1=NC(C2=CC=CC(C)=N2)=C(C3=CC=C4N=CC=NC4=C3)N1)C
Molekylvægt (g/mol) 343.42
Formel vægt 343.42
Procent renhed 95.49%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease
Vis mere Vis mindre

Research purposes only

Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.