Få mere at vide
SB 431542, TRC
Artikelnummer. 30417547
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30417547 10 mg
30423356 25 mg
30425485 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30417547 Leverandør Toronto Research Chemicals Leverandørnr. TRCS15466010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 301836-41-9
Molekylær formel C22 H16 N4 O3
Molekylvægt (g/mol) 384.39
Synonym SB 431542, 4-[4-(1,3-Benzodioxol-5-yl)-5-(2-pyridinyl)-1H-imidazol-2-yl]-benzamide
IUPAC navn 4-[4-(1,3-benzodioxol-5-yl)-5-pyridin-2-yl-1H-imidazol-2-yl]benzamide
SMIL NC(=O)c1ccc(cc1)c2nc(c3ccc4OCOc4c3)c([nH]2)c5ccccn5
Tekniske data
Kemisk navn eller materiale SB 431542
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C22H16N4O3/c23-21(27)13-4-6-14(7-5-13)22-25-19(20(26-22)16-3-1-2-10-24-16)15-8-9-17-18(11-15)29-12-28-17/h1-11H,12H2,(H2,23,27)(H,25,26)
Formel vægt 384.1222
Procent renhed >95%