missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ SB 269970 hydrochloride

Product Code. 18054372
Change view
Click to view available options
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
18054372 10 mg
30001272 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18054372 Supplier Tocris Bioscience™ Supplier No. 1612/10
  • ONLINE EXCLUSIVE PRICE
    2545.00 DKK
    2406.60 DKK / 10mg
    Spare 138.40 DKK
    5% Slukket
 Begrænset vare
Forventet forsendelse: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective 5-HT7 antagonist; brain penetrant

Potent and selective 5-HT7 receptor antagonist (pKi values are 8.9, 7.2 and 6.0 for 5-HT7A, 5-HT5A and 5-HT1B and < 6.0 for 5-HT1A, 5-HT1D, 5-HT1E, 5-HT1F, 5-HT2A, 5-HT2B, 5-HT2C, 5-HT4 and 5-HT6 receptors respectively). Brain penetrant in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261901-57-9
Molekylær formel C18H29ClN2O3S

Specifications

Kemisk navn eller materiale SB 269970 hydrochloride
Mængde 10 mg
Formel vægt 388.95
Renhed >98%
Molekylvægt (g/mol) 388.951
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.