Få mere at vide
Tocris Bioscience™ SB 239063
Beskrivelse
Potent and selective p38 MAP kinase inhibitor (IC50 = 44 nM for p38α). Displays > 220-fold selectivity over ERK, JNK1 and other kinases; ∽ 3-fold more selective than SB 203580 (Cat. Nos. 1202 and 1402). Reduces inflammatory cytokine production and is neuroprotective following oral administration in vivo.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | SB 239063 |
| CAS | 193551-21-2 |
| Mængde | 10 mg |
| Molekylær formel | C20H21FN4O2 |
| Formel vægt | 368.41 |
| Renhed | >98% |
| Molekylvægt (g/mol) | 368.41 |