Få mere at vide
SB-224289 hydrochloride, MedChemExpress
Artikelnummer. 16473268
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16473268 10 mg
16405997 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16473268 Leverandør Medchem Express Leverandørnr. HY101105A
MedChemExpress SB-224289 hydrochloride is a selective 5-HT1B receptor antagonist, with anxiolytic effect.
SB-224289 hydrochloride is a selective 5-HT1B receptor antagonist, with anxiolytic effect.
Tekniske data
Kemisk navn eller materiale SB-224289 hydrochloride
CAS 180084-26-8
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C32H33ClN4O3
Mængde 10 mg
Synonym SB-224289A
Opløselighedsinformation DMSO : 14.29 mg/mL (25.65 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMIL O=C(C1=CC=C(C2=CC=C(C3=NOC(C)=N3)C=C2C)C=C1)N4CCC5=C4C=C6C(OCC67CCN(C)CC7)=C5.Cl
Molekylvægt (g/mol) 557.08
Formel vægt 557.08
Procent renhed 96.4%
Grad Research
Fysisk form Powder
Til brug med (applikation) Neuroscience-Neuromodulation