Få mere at vide
Tocris Bioscience™ SB 224289 hydrochloride
Beskrivelse
SB 224289 hydrochloride is a selective 5-HT1B receptor antagonist (pKi = 8.2). Displays > 60-fold selectivity over 5-HT1D, 5-HT1A, 5-HT1E, 5-HT1F, 5-HT2A and 5-HT2C receptors in radioligand binding and functional assays. Centrally active following oral administration in vivo.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1'-Methyl-5-[[2'-methyl-4'-(5-methyl-1,2,4-oxadiazol-3-yl)biphenyl-4-yl]carbonyl]-2,3,6,7-tetrahydrospiro[furo[2,3-f]indole-3,4'-piperidine hydrochloride |
| CAS | 180084-26-8 |
| Analyseprocentområde | >95% |
| Mængde | 10 mg |
| Opløselighed | Soluble to 10mM with gentle warming in DMSO |
| Molekylær formel | C32H32N4O3.HCl |
| Formel vægt | Observed MW: 557.09 |
For Research Use Only