missing translation for 'onlineSavingsMsg'
Learn More
SB 204990, MedChemExpress Product Code.: 16479597

SB 204990, MedChemExpress

Product Code. p-200102195
5 mg, 5mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16479597

Brand: Medchem Express HY16450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SB 204990 is a potent and specific inhibitor of ATP citrate lyase (ACLY) enzyme.

SB 204990 is a potent and specific inhibitor of ATP citrate lyase (ACLY) enzyme.

Specifications

Chemical Name or Material SB 204990
CAS 154566-12-8
Color White
Purity Grade Notes Research
Molecular Formula C18H22Cl2O5
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (256.89 mM) ∣H2O : 25 mg/mL (64.22 mM; Need ultrasonic)
SMILES O=C(O)C[C@]1(O)C(O[C@H](CCCCCCC2=CC=C(Cl)C=C2Cl)C1)=O
Molecular Weight (g/mol) 389.27
Formula Weight 389.27
Percent Purity 95.35%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
SB 204990, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.